
Diethyl phosphonate, Sigma-Aldrich®
Catalog # SIAL365920-1L
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:138.1 g/mol
Boiling Pt:50 °C (1013 hPa)
Density:1.093 g/cm³ (0 °C)
:M-Clarity™ quality level = MQ200
EINECS:212-091-6
:GHS07
CAS Number:762-04-9
Formula:C₄H₁₁O₃P
MDL Number:MFCD00044573