Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Diethyl phosphonate
Diethyl phosphonate ≥96%
Diethyl phosphonate ≥96%
:
Thermo Fisher Scientific
Diethyl phosphonate ≥96%
:
Thermo Fisher Scientific
:
762-04-9
:
:
C₄H₁₁O₃P
:
MFCD00044573
:
138.1 g/mol
:
50…51 °C (2 mmHg)
:
–70 °C
:
1.070
:
74 °C (165 °F)
:
Ambient
:
212-091-6
:
GHS05
:
GHS07
externalurl.pdf
msds_ALFA043801
Thermo Scientific Acroseal 25 Years