0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Arsenic (III) oxide
Arsenic(III) oxide ≥99.5% (metals basis)
Arsenic(III) oxide ≥99.5% (metals basis)
:
Thermo Fisher Scientific
Arsenic(III) oxide ≥99.5% (metals basis)
:
Thermo Fisher Scientific
:
1327-53-3
:
:
As₂O₃
:
MFCD00003433
:
197.84 g/mol
:
14,804
:
465 °C (subl.)
:
312.3 °C
:
3.74
:
215-481-4
:
1561
:
6.1,II
:
GHS06
:
GHS08
:
GHS05
:
GHS09
:
Ambient
msds_ALFA043488
Thermo Scientific Acroseal 25 Years