

Bismuth(III) nitrate pentahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10035-06-0
:
Formula:Bi(NO₃)₃·5H₂O
MDL Number:MFCD00149157
Boiling Pt:75…80 °C (1013 hPa)
Melting Pt:30 °C
Density:2.736 g/cm³ (20 °C)
ADR:5.1,II
:M-Clarity™ quality level = MQ100
:485.07 g/mol
EINECS:233-791-8
UN:1477
:GHS03
:GHS07