

Bismuth(III) nitrate pentahydrate
Catalog # BIOT000710-0250
Supplier: BIONET RESEARCH
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
Specifications
About this item
:
Boiling Pt:75…80 °C (1013 hPa)
Melting Pt:30 °C
Density:2.736 g/cm³ (20 °C)
ADR:5.1,II
:485.07 g/mol
EINECS:233-791-8
UN:1477
:GHS03
:GHS07
CAS Number:10035-06-0
Formula:Bi(NO₃)₃·5H₂O
MDL Number:MFCD00149157