Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Hexaphenyldisilane
Hexaphenyldisilane
Hexaphenyldisilane
Hexaphenyldisilane
Supplier:
CHEMPUR SILANE
Certificates
Expand image
Hexaphenyldisilane
Supplier:
CHEMPUR SILANE
About this item
CAS Number:
1450-23-3
:
Formula:
C₃₆H₃₀Si₂
MDL Number:
MFCD00039558
:
518.81 g/mol
EINECS:
215-913-1
:
GHS07
Storage Temperature:
Ambient