Hexaphenyldisilane ≥98%
Catalog # L04928.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:518.81 g/mol
Melting Pt:368…370 °C
EINECS:215-913-1
:GHS07
Storage Temperature:Ambient
CAS Number:1450-23-3
Formula:C₃₆H₃₀Si₂
MDL Number:MFCD00039558