Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,1,2,2-Tetraphenyldisilane
1,1,2,2-Tetraphenyldisilane
1,1,2,2-Tetraphenyldisilane
1,1,2,2-Tetraphenyldisilane
Supplier:
CHEMPUR SILANE
Certificates
Expand image
1,1,2,2-Tetraphenyldisilane
Supplier:
CHEMPUR SILANE
About this item
CAS Number:
16343-18-3
:
Formula:
C₂₄H₂₂Si₂
MDL Number:
MFCD00093170
:
366.61 g/mol