Chlorotriphenylsilane
Supplier: CHEMPUR SILANE
Certificates
About this item
CAS Number:76-86-8
:
Formula:C₁₈H₁₅ClSi
MDL Number:MFCD00000496
:294.86 g/mol
Boiling Pt:378 °C (1013 hPa)
Melting Pt:90…95 °C
Density:1.0165 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-989-0
UN:3261
ADR:8,II
:GHS05