Chlorotriphenylsilane 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:76-86-8
:
Formula:C₁₈H₁₅ClSi
MDL Number:MFCD00000496
:294.86 g/mol
Boiling Pt:188…189 °C (3 mmHg)
Melting Pt:91…96 °C
Density:1.0165 g/cm³ (20 °C)
Flash Pt:>200 °C (392 °F)
Storage Temperature:Ambient
EINECS:200-989-0
UN:3261
ADR:8,II
:GHS05