Chlorotriphenylsilane 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:76-86-8
:
Formula:C₁₈H₁₅ClSi
MDL Number:MFCD00000496
:294.86 g/mol
Boiling Pt:378 °C (1013 hPa)
Melting Pt:91…96 °C
Density:1,0165 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-989-0
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to beige Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:91 to 96 °C
- GC:≥94 %