Diphenylsilane
Supplier: CHEMPUR SILANE
Certificates
About this item
CAS Number:775-12-2
:
Formula:C₁₂H₁₂Si
MDL Number:MFCD00003002
:184.31 g/mol
Boiling Pt:95 °C (1013 hPa)
Density:0,9962 g/cm³ (20 °C)
EINECS:212-271-4
:GHS07
Storage Temperature:Ambient