Tetrapropyltin ≥97%
Catalog # STRE93-5053-1G
Supplier: Strem Chemicals, Inc.
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:291.06 g/mol
Boiling Pt:222 °C
Density:1.1065 (20°)
EINECS:218-536-0
UN:2788
ADR:6.1,II
:GHS07
:GHS09
CAS Number:2176-98-9
Formula:C₁₂H₂₈Sn
MDL Number:MFCD00015196