Order Entry
Germany
ContactUsLinkComponent
Tetrapropyltin
Tetrapropyltin
Catalog # PROODE-MS-02122-03
CAS Number:  
Tetrapropyltin
Catalog # PROODE-MS-02122-03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    300 mg
  • Seq:
    2625

Specifications

About this item

 :
 :291.06 g/mol
EINECS:218-536-0
UN:2788
ADR:6.1,II
 :GHS06
 :GHS09
CAS Number:2176-98-9
Formula:C₁₂H₂₈Sn
MDL Number:MFCD00015196