Tetrapropyltin
Catalog # EHERC17415700
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:291.06 g/mol
EINECS:218-536-0
UN:2788
ADR:6.1,II
:GHS06
:GHS09
CAS Number:2176-98-9
Formula:C₁₂H₂₈Sn
MDL Number:MFCD00015196