Chlorotrihexylsilane, Sigma-Aldrich®
Catalog # SIAL254444-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:319.05 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:222-851-9
UN:2987
:GHS05
CAS Number:3634-67-1
Formula:C₁₈H₃₉ClSi
MDL Number:MFCD00000510