Order Entry
Germany
ContactUsLinkComponent
Triisopropylchlorosilane 95%
Triisopropylchlorosilane 95%
Certificates
undefined
Triisopropylchlorosilane 95%

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4520 to 1.4540 (20°C, 589 nm)
  • GC:
    ≥94 %

Specification Test Results

Related Information