Triisopropylchlorosilane
Catalog # ABCRAN111480.0100
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
About this item
:
:192.8 g/mol
Boiling Pt:198 °C (1013 hPa)
Density:0.9008 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2987
ADR:8,II
:GHS05
CAS Number:13154-24-0
Formula:C₉H₂₁ClSi
MDL Number:MFCD00009656