(3-Mercaptopropyl)trimethoxysilane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:4420-74-0
:
Formula:HSH₂CCH₂CH₂Si(OCH₃)₃
MDL Number:MFCD00004901
:196.34 g/mol
Boiling Pt:215 °C (1013 hPa)
Melting Pt:–50 °C
Density:1.056 g/cm³ (20 °C)
Flash Pt:>65 °C
Storage Temperature:Ambient
UN:3082
ADR:9,III
:M-Clarity™ quality level = MQ200
EINECS:224-588-5