Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Propyltrimethoxysilane
Propyltrimethoxysilane
Propyltrimethoxysilane
Propyltrimethoxysilane
Supplier:
CHEMPUR SILANE
Certificates
Expand image
Propyltrimethoxysilane
Supplier:
CHEMPUR SILANE
About this item
CAS Number:
1067-25-0
:
Formula:
C₆H₁₆O₃Si
MDL Number:
MFCD00043026
:
164.28 g/mol
Boiling Pt:
142…143 °C (1013 hPa)
Density:
0,935 g/cm³ (20 °C)
EINECS:
213-926-7