Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Octyltrimethoxysilane
Octyltrimethoxysilane
Octyltrimethoxysilane
Octyltrimethoxysilane
Supplier:
CHEMPUR SILANE
Certificates
Expand image
Octyltrimethoxysilane
Supplier:
CHEMPUR SILANE
About this item
CAS Number:
3069-40-7
:
Formula:
C₁₁H₂₆O₃Si
MDL Number:
MFCD00043053
:
234.41 g/mol
EINECS:
221-338-7