Octyltriethoxysilane, Sigma-Aldrich®
Catalog # SIAL679305-50G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:276.49 g/mol
Boiling Pt:265 °C (1013 hPa)
Melting Pt:–40 °C
Density:0.8515 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:220-941-2
:GHS07
CAS Number:2943-75-1
Formula:C₁₄H₃₂O₃Si
MDL Number:MFCD00039883