Octyltriethoxysilane 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2943-75-1
:
Formula:C₁₄H₃₂O₃Si
MDL Number:MFCD00039883
:276.49 g/mol
Boiling Pt:265 °C
Melting Pt:< –40 °C
Density:0.878
Flash Pt:109 °C (228 °F)
EINECS:220-941-2
:GHS07
Storage Temperature:Ambient