(3-Glycidoxypropyl)trimethoxysilane
Supplier: CHEMPUR SILANE
Certificates
About this item
CAS Number:2530-83-8
:
Formula:C₉H₂₀O₅Si
MDL Number:MFCD00005144
:236.34 g/mol
Boiling Pt:299 °C (1013 hPa)
Melting Pt:0 °C
Density:1.07 g/cm³ (20 °C)
Flash Pt:122 °C
Storage Temperature:Ambient
EINECS:219-784-2