


Lead(II) sulphide ≥99.995% (metals basis)
Catalog # 45462.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
CAS Number:1314-87-0
:
Formula:PbS
MDL Number:MFCD00016280
:239.27 g/mol
Merck Index:14,5421
Boiling Pt:1344 °C (1013 hPa)
Melting Pt:1114 °C
Density:7.5
Storage Temperature:Ambient
EINECS:215-246-6
UN:3077
ADR:9,III
:GHS08
:GHS07
:GHS09
Formula:̍PbS