Triethoxysilane
Marketsource
Supplier: CHEMPUR SILANE
Jump to:
About this item
- CAS Number:998-30-1
- Formula:C₆H₁₆O₃Si
- MDL Number:MFCD00009061
- Molecular Weight:164.28 g/mol
- Boiling Pt:135 °C (1013 hPa)
- Melting Pt:–170 °C
- Density:0.8753 g/cm³ (20 °C)
- EINECS:213-650-7
- UN:3384
- ADR:6.1,I
3 Options Available Below
Product Details & Documents
- CAS Number:998-30-1
- Formula:C₆H₁₆O₃Si
- MDL Number:MFCD00009061
- Molecular Weight:164.28 g/mol
- Boiling Pt:135 °C (1013 hPa)
- Melting Pt:–170 °C
- Density:0.8753 g/cm³ (20 °C)
- EINECS:213-650-7
- UN:3384
- ADR:6.1,I
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



