0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Germanium (IV) oxide
Germanium(IV) oxide ≥99.9999% (metals basis)
Germanium(IV) oxide ≥99.9999% (metals basis)
:
Thermo Fisher Scientific
Germanium(IV) oxide ≥99.9999% (metals basis)
:
Thermo Fisher Scientific
:
1310-53-8
:
:
GeO₂
:
MFCD00011030
:
104.61 g/mol
:
14,4409
:
1086 °C
:
6.239
:
215-180-8
:
GHS07
:
Ambient
msds_ALFA010510
Thermo Scientific Acroseal 25 Years