Tetrafluorosilane
Catalog # SILNSILS15525.25
Supplier: CHEMPUR SILANE
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:104.08 g/mol
Merck Index:12,08645
Boiling Pt:–86 °C (1013 hPa)
Melting Pt:–77 °C
Density:0.00469 g/cm³ (20 °C)
EINECS:232-015-5
UN:1859
ADR:2,
CAS Number:7783-61-1
Formula:SiF₄