


Lead(II) oxide ≥99.9995% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1317-36-8
:
Formula:PbO
MDL Number:MFCD00011164
:223.2 g/mol
Merck Index:14,5413
Boiling Pt:1470 °C (1013 hPa)
Melting Pt:886 °C
Density:9.53
Storage Temperature:Ambient
EINECS:215-267-0
UN:3077
ADR:9,III
:GHS08
:GHS07
:GHS09
Specification Test Results
- Total Metal Impurities:0,0005% max.
Specification Test Results
Frequently Bought Together