
Silicon carbide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:409-21-2
:
Formula:SiC
MDL Number:MFCD00049531
:40.1 g/mol
Melting Pt:2700 °C
Density:3.22 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ100
EINECS:206-991-8
:GHS07