Order Entry
Germany
ContactUsLinkComponent
Silicon carbide ≥99.95% (metals basis excluding B), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Irritant
Silicon carbide ≥99.95% (metals basis excluding B), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 41084.KS
CAS Number:  
undefined
Irritant
Silicon carbide ≥99.95% (metals basis excluding B), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 41084.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:409-21-2
 :
Formula:SiC
MDL Number:MFCD00049531
 :40.1 g/mol
Melting Pt:2700 °C
Density:3.22 g/cm³ (25 °C)
EINECS:206-991-8
 :GHS07
Storage Temperature:Ambient