


Lead(II) bromide ≥99.998% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10031-22-8
:
Formula:Br₂Pb
MDL Number:MFCD00011156
:367.01 g/mol
Merck Index:14,5403
Boiling Pt:916 °C
Melting Pt:373 °C
Density:6.66
Storage Temperature:Ambient
EINECS:233-084-4
UN:2291
ADR:6.1,III
:GHS08
:GHS07
:GHS09