Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
di-Boron trioxide
Diboron trioxide, Supelco®
Diboron trioxide, Supelco®
:
Merck
Diboron trioxide, Supelco®
:
Merck
:
1303-86-2
:
:
B₂O₃
:
MFCD00011315
:
69.62 g/mol
:
13,01327
:
1860 °C (1013 hPa)
:
450 °C
:
1.81…1.86 g/cm³ (20 °C)
:
Ambient
:
215-125-8
:
01-2119486655-24
:
GHS08