Order Entry
Germany
ContactUsLinkComponent
 
Aluminium oxide, powder, BAKER ANALYZED® for chromatography, J.T.Baker®
  0537-01
 :  Avantor
 :  
undefined
Aluminium oxide, powder, BAKER ANALYZED® for chromatography, J.T.Baker®
  0537-01
 :  Avantor
 :  

 

  • Pack type:
    Plastic bottle, T/E cap
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • For Laboratory,Research,or Manufacturing Use:
    7.0 - 7.8
  • pH of 5% Slurry at 20°C:
    6.0 - 7.5
  • Adsorption, mg o-nitroaniline/g:
    ≥0.05
  • Loss on Drying:
    ≤5 %

 

 

 :
 :101.96 g/mol
 :13,00355
 :2980 °C (1013 hPa)
 :2045 °C
 :3.94 g/cm³ (20 °C)
 :215-691-6
 :01-2119529248-35
 :1344-28-1
 :11092-32-3
 :Al₂O₃
 :MFCD00003424