Indium(III) chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10025-82-8
:
Formula:Cl₃In
MDL Number:MFCD00011058
:221.18 g/mol
Merck Index:13,04980
Boiling Pt:800 °C (1013 hPa)
Melting Pt:586 °C
Density:4 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3260
ADR:8,II
:M-Clarity™ quality level = MQ100
EINECS:233-043-0