Barium sulphate, fine powder Ph. Eur.
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7727-43-7
:
Formula:BaSO₄
MDL Number:MFCD00003455
:233.39 g/mol
Merck Index:13,00997
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:1345 °C
Density:4.5 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-784-4
Specification Test Results
- Appearance:White powder
- Identification A:Passes test
- Identification B:Passes test
- Acidity or alkalinity:Passes test
- Substances soluble in acid:Max. 0.3 %
- Oxidisable sulphur compounds:Passes test
- Soluble barium salts:Max. 10 ppm
- Loss on ignition (600 ± 50°C):Max. 2 %
- Residual solvents:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number2,5L Plastic bottle for solids DIN 80 (ZPACK13252500) 50L Plastic bucket UN Y (ZPACK13247050)
Packaging Information
spec_VWRC21763.292 spec_VWRC21763.292 spec_VWRC21763.463 spec_VWRC21763.463
Product specification
VWR Chemicals for Biopharma and Pharma Production