Calcium fluoride
25 kg
Catalog # HONE01124.9025
Supplier: Honeywell Chemicals
Jump to:
Product Details & Documents
- CAS Number:7789-75-5
- Formula:CaF₂
- MDL Number:MFCD00010907
- Molecular Weight:78.07 g/mol
- Merck Index:13,01669
- Boiling Pt:2500 °C (1013 hPa)
- Melting Pt:1418 °C
- Density:3.18 g/cm³ (20 °C)
- EINECS:232-188-7
Specifications
Specifications
- Pk:25 kg



