Strontium carbonate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1633-05-2
:
Formula:CO₃Sr
MDL Number:MFCD00011250
Merck Index:14,08838
Boiling Pt:2647 °C (1 mmHg)
Melting Pt:1768.85 °C
Density:3.548 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:147.63 g/mol
Storage Temperature:Ambient
EINECS:216-643-7