Strontium carbonate
Catalog # HONE13904.9025
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pk:25 kg
- Seq:4283
Specifications
About this item
:
Merck Index:14,08838
Boiling Pt:2647 °C (1 mmHg)
Melting Pt:1768.85 °C
Density:3.548 g/cm³ (20 °C)
:147.63 g/mol
Storage Temperature:Ambient
EINECS:216-643-7
CAS Number:1633-05-2
Formula:CO₃Sr
MDL Number:MFCD00011250