

Barium perchlorate, EMSURE® for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:13465-95-7
:
Formula:Ba(ClO₄)₂
MDL Number:MFCD00003440
:336.23 g/mol
Merck Index:12,01016
Melting Pt:505 °C
Density:3.2 g/cm³ (20 °C)
Flash Pt:21 °C (closed cup)
UN:1447
ADR:5.1 (6.1),II
:M-Clarity™ quality level = MQ300
EINECS:236-710-4
:GHS03
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:1985 g/l (25 °C)
- Melting Point:505 °C
- Molar Mass:336.23 g/mol
- Bulk Density:1300 kg/m3
- Density:2.74 g/cm3 (20 °C)
- pH:5.6 (100 g/l, H2O, 20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer