Order Entry
Germany
ContactUsLinkComponent
 
 
Calcium chloride, anhydrous ≥93.0%, granular ≤7.0 mm, Fluka™
 
 
Calcium chloride, anhydrous ≥93.0%, granular ≤7.0 mm, Fluka™
 

 

  • Form:
    granular
  • Assay:
    Min. 93.0 %
  • Colour of the solution:
    complying
  • Clearness of the solution:
    complying
  • Granulation:
    complying

 

 

Frequently Bought Together