
Strontium bromide, Sigma-Aldrich®
Catalog # SIAL430684-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:247.43 g/mol
Merck Index:13,08917
Boiling Pt:2146 °C (1013 hPa)
Melting Pt:643 °C
Density:4.175 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ100
EINECS:233-969-5
:GHS07
CAS Number:10476-81-0
Formula:Br₂Sr
MDL Number:MFCD00049548