Order Entry
Germany
ContactUsLinkComponent
Sodium sulphate, Sigma-Aldrich®
Sodium sulphate, Sigma-Aldrich®
Catalog # SIALS5640-500G
Supplier:  Merck
CAS Number:  
undefined
Sodium sulphate, Sigma-Aldrich®
Catalog # SIALS5640-500G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
Merck Index:12,08829
Boiling Pt:1429 °C (1013 hPa)
Melting Pt:888 °C
Density:2.7 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
 :142.04 g/mol
Storage Temperature:Ambient
EINECS:231-820-9
CAS Number:7757-82-6
Formula:Na₂SO₄
MDL Number:MFCD00003504