Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium selenite
Sodium selenite, Sigma-Aldrich®
Sodium selenite, Sigma-Aldrich®
:
Merck
Sodium selenite, Sigma-Aldrich®
:
Merck
:
10102-18-8
:
:
Na₂SeO₃
:
MFCD00003489
:
12,08822
:
710 °C
:
3.1 g/cm³ (20 °C)
:
Ambient
:
6.1,I
:
M-Clarity™ quality level = MQ300
:
172.94 g/mol
:
233-267-9
:
2630
:
GHS06
:
GHS09