Order Entry
Germany
ContactUsLinkComponent
Potassium disulphite, EMSURE® for analysis, Supelco®
Potassium disulphite, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates
Potassium disulphite, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    450 g/l (20 °C)
  • Melting Point:
    150 °C (decomposition)
  • Molar Mass:
    222.33 g/mol
  • Bulk Density:
    1000 - 1300 kg/m3
  • Density:
    2.34 g/cm3
  • pH:
    3.0 - 4.5 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number