

Potassium disulphite, extra pure
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:16731-55-8
:
Formula:K₂O₅S₂
MDL Number:MFCD00167605
:222.33 g/mol
Merck Index:13,07729
Melting Pt:190 °C
Density:1.2 g/cm³ (20 °C)
EINECS:240-795-3
:GHS05
:GHS07