Order Entry
Germany
ContactUsLinkComponent
Potassium disulphite ≥96% (by iodometry), powder, Puriss. p.a., Fluka™
Potassium disulphite ≥96% (by iodometry), powder, Puriss. p.a., Fluka™
Certificates
Potassium disulphite ≥96% (by iodometry), powder, Puriss. p.a., Fluka™
Certificates

Specification Test Results

  • Assay (Iodometric):
    96 - 101.0 %
  • Arsenic (As):
    Max. 0.0001 %
  • Copper (Cu):
    Max. 0.001 %
  • Iron (Fe):
    Max. 0.0005 %
  • Lead (Pb):
    Max. 0.0005 %
  • Zinc (Zn):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 0.005 %

Specification Test Results

Related Information