0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
di-Potassium disulphite
Potassium disulphite ≥95%
Potassium disulphite ≥95%
:
Thermo Fisher Scientific
Potassium disulphite ≥95%
:
Thermo Fisher Scientific
:
16731-55-8
:
:
K₂O₅S₂
:
MFCD00167605
:
222.33 g/mol
:
14,7645
:
190 °C (dec.)
:
1.2 g/cm³ (20 °C)
:
240-795-3
:
GHS07
:
Ambient
msds_ALFA043908
Thermo Scientific Acroseal 25 Years