Potassium hexachloroplatinate(IV), Sigma-Aldrich®
: Merck
:16921-30-5
:
:K₂PtCl₆
:MFCD00011389
:13,07720
:250 °C
:3.344 g/cm³ (20 °C)
:Ambient
:6.1,II
:M-Clarity™ quality level = MQ100
:485.99 g/mol
:240-979-3
:3290
:GHS06
:GHS05
:GHS08