Sodium dihydrogen phosphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7558-80-7
:
Formula:NaH₂PO₄
MDL Number:MFCD00003527
Merck Index:12,08806
Melting Pt:∼650 °C
Density:0.9996 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
:119.98 g/mol
Storage Temperature:Ambient
EINECS:231-449-2
REACH:01-2119489796-13